Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100364958 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253861.2 | XP_006253923.1 | 2 | 234 |
Gene Symbol: Tubb2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109119.1 | NP_001102589.1 | 258 | |||
XM_008771582.1 | XP_008769804.1 | 283 |
Gene Symbol: Tubb2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X03369.1 | - | 3 | 189 |