Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lancl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014187.1 | NP_001014209.1 | 4, 5 | 693 | ||
XM_006236580.2 | XP_006236642.2 | 4, 5 | 555 | |||
XM_006236581.2 | XP_006236643.1 | 4, 5 | 702 | |||
GenBank mRNA | BC079412.1 | - | 4, 5 | 693 | 7113320 |