Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other C1ORF109 SIRNA Assays ›
Gene Symbol:
C1orf109
Gene Name:
chromosome 1 open reading frame 109
Gene Aliases:
-
Chromosome Location:
Chr.1: 37681570 - 37692336 on Build GRCh38
UniGene ID:
Hs.272673
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
25598
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: C1orf109
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001303030.1
NP_001289959.1
3
820
NM_001303031.1
NP_001289960.1
3
823
NM_017850.2
NP_060320.1
3
702
XM_005270975.4
XP_005271032.1
3
1675
XM_005270979.1
XP_005271036.1
3
663
XM_005270980.1
XP_005271037.1
3
639
XM_005270981.4
XP_005271038.1
3
1889
XM_005270982.1
XP_005271039.1
3
971
XM_011541637.1
XP_011539939.1
5
1096
XM_011541638.1
XP_011539940.1
4
973
XM_011541639.1
XP_011539941.1
4
970
XM_011541640.1
XP_011539942.1
5
975
XM_011541641.1
XP_011539943.1
4
852
XM_011541642.1
XP_011539944.1
4
855
XM_011541643.1
XP_011539945.1
4
873
XM_011541646.2
XP_011539948.1
3
1678
XM_017001544.1
XP_016857033.1
4
1065
XM_017001545.1
XP_016857034.1
5
829
XM_017001546.1
XP_016857035.1
3
1888
XM_017001547.1
XP_016857036.1
3
971
XM_017001548.1
XP_016857037.1
5
1093
XM_017001549.1
XP_016857038.1
4
970
XM_017001550.1
XP_016857039.1
4
973
XM_017001551.1
XP_016857040.1
3
639
XM_017001552.1
XP_016857041.1
5
975
XM_017001553.1
XP_016857042.1
5
837
XM_017001554.1
XP_016857043.1
4
852
XM_017001555.1
XP_016857044.1
4
855
XM_017001556.1
XP_016857045.1
3
1318
GenBank mRNA
AK000515.1
-
3
627
AK299377.1
-
3
719
AK316093.1
-
3
676
BC018109.1
-
5
803
3914920
BC063843.1
-
3
728
6501461
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline