Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109110.1 | NP_001102580.1 | 1097 |
Gene Symbol: Palld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC088409.1 | - | 4 | 1236 | 7189104 |