Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014254.1 | NP_001014276.1 | 7 | 959 | ||
XM_008761185.1 | XP_008759407.1 | 7 | 1164 | |||
XM_008761186.1 | XP_008759408.1 | 8 | 1031 | |||
XM_008761187.1 | XP_008759409.1 | 7 | 1161 | |||
XM_008761188.1 | XP_008759410.1 | 7 | 1143 | |||
XM_008761189.1 | XP_008759411.1 | 7 | 1140 | |||
XM_008761190.1 | XP_008759412.1 | 7 | 1140 | |||
XM_008761191.1 | XP_008759413.1 | 7 | 1137 | |||
XM_008761192.1 | XP_008759414.1 | 7 | 1010 | |||
XM_008761193.1 | XP_008759415.1 | 4 | 811 | |||
XM_008761194.1 | XP_008759416.1 | 4 | 290 | |||
GenBank mRNA | BC083788.1 | - | 7 | 959 | 7098251 |