Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024276.1 | NP_001019447.1 | 2 | 883 | ||
GenBank mRNA | BC092587.1 | - | 2 | 876 | 7368213 | |
BC092590.1 | - | 2 | 883 | 7371843 |