Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sufu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024899.1 | NP_001020070.1 | 4, 5 | 761 | ||
XM_006231546.2 | XP_006231608.1 | 4, 5 | 815 | |||
XM_008760443.1 | XP_008758665.1 | 3, 4 | 821 | |||
GenBank mRNA | BC097447.1 | - | 4, 5 | 761 | 7456556 |