Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008096.2 | NP_032122.1 | 13 | 1745 | ||
GenBank mRNA | AK010965.1 | - | 13 | 1569 | ||
BC010762.1 | - | 13 | 1608 | 4234788 | ||
BC051395.1 | - | 13 | 1612 | 5125043 | ||
M55413.1 | - | 13 | 1555 |