Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ly6g6e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027366.1 | NP_081642.1 | 3 | 1021 | ||
GenBank mRNA | AK009303.1 | - | 3 | 878 | ||
BB397904.1 | - | 1 | 206 |