Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166269.1 | NP_001159741.1 | 4, 5 | 578 | ||
NM_017815.2 | NP_060285.2 | 4, 5 | 570 | |||
GenBank mRNA | AK000431.1 | - | 4, 5 | 539 | ||
AK026016.1 | - | 4, 5 | 524 | |||
BC001916.1 | - | 4, 5 | 567 | 2905474 | ||
BC002554.2 | - | 4, 5 | 501 | 3139198 | ||
BQ066232.1 | - | 4, 5 | 578 | |||
BX247995.1 | - | 3, 4 | 387 |