Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR1J2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054107.1 | NP_473448.1 | 1 | 256 | ||
XM_017014623.1 | XP_016870112.1 | 10 | 1706 | |||
GenBank mRNA | AK123016.1 | - | 4 | 582 |