Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNL4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142317.1 | NP_001135789.1 | 4 | 1800 | ||
NM_001142318.1 | NP_001135790.1 | 4 | 1698 | |||
NM_001324355.1 | NP_001311284.1 | 5 | 1949 | |||
NM_017853.2 | NP_060323.1 | 4 | 1849 | |||
GenBank mRNA | AK000518.1 | - | 4 | 1823 | ||
BC009646.1 | - | 4 | 1791 | 3458545 |