Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166304.1 | NP_001159776.1 | 7 | 1059 | ||
NM_017861.3 | NP_060331.3 | 6 | 1005 | |||
GenBank mRNA | AK294340.1 | - | 8 | 1359 | ||
BC022542.1 | - | 6 | 965 | 4797783 |