Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012417.2 | NP_001012417.2 | 3 | 419 | ||
NM_001100909.1 | NP_001094379.1 | 3 | 419 | |||
NM_001278320.1 | NP_001265249.1 | 3 | 419 | |||
GenBank mRNA | AY324188.1 | - | 3 | 419 | ||
AY324189.1 | - | 3 | 419 | |||
AY324190.1 | - | 3 | 419 | |||
BC140799.1 | - | 3 | 293 | 9021667 | ||
BC171746.1 | - | 2 | 654 |