Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318533.1 | NP_001305462.1 | 6 | 471 | ||
NM_017868.3 | NP_060338.3 | 6 | 471 | |||
XM_005271604.3 | XP_005271661.1 | 6 | 687 | |||
XM_011542890.2 | XP_011541192.1 | 6 | 518 | |||
XM_017017943.1 | XP_016873432.1 | 6 | 471 | |||
XM_017017944.1 | XP_016873433.1 | 7 | 1041 | |||
XM_017017945.1 | XP_016873434.1 | 7 | 524 | |||
XM_017017946.1 | XP_016873435.1 | 6 | 694 | |||
XM_017017947.1 | XP_016873436.1 | 6 | 471 | |||
XM_017017948.1 | XP_016873437.1 | 6 | 471 | |||
XM_017017949.1 | XP_016873438.1 | 6 | 471 | |||
XM_017017950.1 | XP_016873439.1 | 6 | 471 | |||
GenBank mRNA | AK000542.1 | - | 5 | 363 | ||
AK023921.1 | - | 6 | 417 | |||
AK091887.1 | - | 7 | 916 | |||
AK223117.1 | - | 5 | 363 | |||
AK292331.1 | - | 6 | 409 | |||
BC011546.1 | - | 5 | 395 | |||
BC032355.1 | - | 5 | 393 | 5219117 | ||
DA954030.1 | - | 6 | 472 | |||
DB110945.1 | - | 6 | 471 |