Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vasp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282021.1 | NP_001268950.1 | 2 | 478 | ||
NM_001282022.1 | NP_001268951.1 | 2 | 478 | |||
NM_009499.3 | NP_033525.2 | 2 | 478 | |||
GenBank mRNA | AF084548.1 | - | 2 | 445 | ||
AK140329.1 | - | 2 | 442 | |||
AK150103.1 | - | 2 | 440 | |||
AK155480.1 | - | 2 | 440 | |||
AK157021.1 | - | 2 | 442 | |||
AK170780.1 | - | 2 | 434 | |||
AK171302.1 | - | 2 | 442 | |||
AK171715.1 | - | 2 | 442 | |||
BC010223.1 | - | 2 | 180 | 3500587 | ||
BC015289.1 | - | 2 | 421 | 4240907 |