Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181751.2 | NP_861416.1 | 1 | 257 | ||
GenBank mRNA | AB233291.1 | - | 1 | 257 | ||
AY288423.1 | - | 1 | 37 | |||
BC115557.1 | - | 1 | 37 | 40101841 | ||
BC115558.1 | - | 1 | 37 | 40101842 |
Set Membership: |
![]() |