Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC642423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_049748.1 | 1 | 570 | |||
GenBank mRNA | AK308544.1 | - | 4 | 925 |
Gene Symbol: GOLGA2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026811.3 | 8 | 1849 | |||
NR_033936.3 | 7 | 2092 | ||||
NR_103496.2 | 7 | 1710 | ||||
GenBank mRNA | AF316855.1 | - | 2 | 1510 | ||
BC007811.2 | - | 9 | 1773 | |||
BC065260.1 | - | 8 | 1766 | |||
BC072390.1 | - | 7 | 2045 |
Gene Symbol: GOLGA2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027001.1 | 7 | 2084 | |||
GenBank mRNA | AK074778.1 | - | 7 | 1717 | ||
BC054509.1 | - | 5 | 3407 | |||
BC071951.1 | - | 7 | 2041 | |||
BC096759.1 | - | 7 | 2041 |
Gene Symbol: LOC727751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102747.1 | 6 | 1905 | |||
NR_102748.1 | 6 | 1610 |