Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318485.1 | NP_001305414.1 | 2 | 205 | ||
NM_017971.3 | NP_060441.2 | 2 | 205 | |||
GenBank mRNA | AB049644.1 | - | 2 | 136 | ||
AK000886.1 | - | 2 | 139 | |||
AK301440.1 | - | 2 | 185 | |||
AK315794.1 | - | 2 | 139 | |||
BC009515.1 | - | 2 | 173 | 3542715 | ||
BC014316.1 | - | 2 | 142 | 4045076 | ||
BC059945.1 | - | 2 | 123 | 5397911 | ||
BG700526.1 | - | 2 | 206 | |||
DN990876.1 | - | 2 | 177 |