Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019287.2 | NP_062160.2 | 3 | 292 | ||
GenBank mRNA | CB801343.1 | - | 3 | 284 |