Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tro | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002272.3 | NP_001002272.1 | 3 | 506 | ||
NM_001290770.1 | NP_001277699.1 | 3 | 455 | |||
NM_001290771.1 | NP_001277700.1 | 3 | 506 | |||
NM_001290772.1 | NP_001277701.1 | 3 | 506 | |||
NM_019548.4 | NP_062421.2 | 3 | 506 | |||
GenBank mRNA | AB032477.1 | - | 2 | 394 | ||
AB036783.1 | - | 3 | 469 | |||
AF241244.1 | - | 2 | 391 | |||
AF241245.1 | - | 2 | 391 | |||
AF288605.1 | - | 3 | 469 | |||
AF288606.1 | - | 3 | 469 | |||
AF319977.1 | - | 2 | 389 | |||
AK047777.1 | - | 3 | 530 | |||
BC075630.1 | - | 3 | 455 | 30536880 | ||
BM938171.1 | - | 2 | 362 |