Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyx1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007010.1 | NP_001007011.1 | 4, 5 | 729 | ||
XM_006243328.2 | XP_006243390.1 | 4, 5 | 809 | |||
XM_006243329.2 | XP_006243391.1 | 4, 5 | 809 | |||
XM_008766313.1 | XP_008764535.1 | 4, 5 | 707 | |||
XM_008766314.1 | XP_008764536.1 | 4, 5 | 538 | |||
GenBank mRNA | AY428506.1 | - | 4, 5 | 729 | ||
BC085838.1 | - | 4, 5 | 673 | 7135744 |