Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007678.2 | NP_001007679.1 | 2 | 1399 | ||
GenBank mRNA | BC161807.1 | - | 2 | 1407 | 9036685 | |
DY319223.1 | - | 1 | 391 |
Gene Symbol: RGD1563962 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749301.3 | XP_003749349.1 | 2 | 1393 |