Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slpil2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008872.1 | NP_001008872.1 | 3 | 365 | ||
XM_008762532.1 | XP_008760754.1 | 3 | 365 |
Gene Symbol: LOC103694893 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773874.1 | XP_008772096.1 | 3 | 365 |