Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010848.3 | NP_001010848.2 | 8, 9 | 1617 | ||
NM_001165972.1 | NP_001159444.1 | 8, 9 | 1614 | |||
XM_011539175.2 | XP_011537477.1 | 8, 9 | 1623 | |||
XM_017015577.1 | XP_016871066.1 | 9, 10 | 1703 | |||
XM_017015578.1 | XP_016871067.1 | 9, 10 | 1698 | |||
XM_017015579.1 | XP_016871068.1 | 9, 10 | 1699 | |||
XM_017015580.1 | XP_016871069.1 | 9, 10 | 1695 | |||
XM_017015581.1 | XP_016871070.1 | 8, 9 | 1620 | |||
XM_017015583.1 | XP_016871072.1 | 9, 10 | 970 | |||
GenBank mRNA | AK122886.1 | - | 10, 11 | 1456 | ||
AK296563.1 | - | 8, 9 | 1439 | |||
BC136811.1 | - | 8, 9 | 1614 | 9020801 | ||
BC143665.1 | - | 6, 7 | 651 | |||
DQ001411.1 | - | 8, 9 | 1590 | |||
HM068874.1 | - | 6, 7 | 1360 | |||
HM068875.1 | - | 9, 10 | 927 | |||
HM068879.1 | - | 8, 9 | 827 | |||
HM068880.1 | - | 8, 9 | 902 | |||
HM068883.1 | - | 9, 10 | 1002 | |||
HM068884.1 | - | 10, 11 | 1080 | |||
HM068885.1 | - | 6, 7 | 673 |