Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014974.1 | NP_001014974.1 | 3 | 1070 | ||
GenBank mRNA | AM690747.1 | - | 1 | 673 | ||
BC085151.1 | - | 3 | 1070 | 30534216 |