Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDH19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105243.1 | NP_001098713.1 | 5 | 8664 | ||
NM_001184880.1 | NP_001171809.1 | 6 | 8805 | |||
NM_020766.2 | NP_065817.2 | 5 | 8661 | |||
XM_011530997.2 | XP_011529299.1 | 6 | 7771 | |||
GenBank mRNA | AB037734.1 | - | 5 | 4869 | ||
AK055951.1 | - | 1 | 1210 | |||
CR749278.1 | - | 5 | 5542 | |||
EF676096.1 | - | 6 | 8805 |