Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZC3H14 SIRNA Assays ›
Gene Symbol:
ZC3H14
Gene Name:
zinc finger CCCH-type containing 14
Gene Aliases:
MSUT-2, NY-REN-37, SUT2, UKp68
Chromosome Location:
Chr.14: 88562909 - 88614720 on Build GRCh38
UniGene ID:
Hs.684046
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
258617
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZC3H14
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001160103.1
NP_001153575.1
17
3652
NM_001160104.1
NP_001153576.1
17
3637
NM_001326295.1
NP_001313224.1
15
3262
NM_001326296.1
NP_001313225.1
16
3577
NM_001326297.1
NP_001313226.1
17
3836
NM_001326298.1
NP_001313227.1
14
3187
NM_001326299.1
NP_001313228.1
16
3562
NM_001326300.1
NP_001313229.1
16
3540
NM_001326301.1
NP_001313230.1
16
3761
NM_001326302.1
NP_001313231.1
16
3537
NM_001326303.1
NP_001313232.1
14
3365
NM_001326304.1
NP_001313233.1
14
3144
NM_001326305.1
NP_001313234.1
15
3465
NM_001326306.1
NP_001313235.1
13
3028
NM_001326307.1
NP_001313236.1
16
3580
NM_001326308.1
NP_001313237.1
14
3147
NM_001326309.1
NP_001313238.1
13
3072
NM_001326310.1
NP_001313239.1
17
3640
NM_001326311.1
NP_001313240.1
13
3069
NM_001326312.1
NP_001313241.1
16
3565
NM_001326313.1
NP_001313242.1
13
3031
NM_001326314.1
NP_001313243.1
15
3462
NM_001326315.1
NP_001313244.1
17
3833
NM_001326316.1
NP_001313245.1
17
3500
NM_024824.4
NP_079100.2
17
3655
NM_207660.3
NP_997543.1
14
3184
NM_207661.2
NP_997544.1
15
3110
NM_207662.3
NP_997545.2
7
2481
NR_136936.1
16
3495
XM_005268068.4
XP_005268125.1
16
3575
XM_005268069.4
XP_005268126.1
15
3257
XM_005268070.4
XP_005268127.1
15
3254
XM_005268071.4
XP_005268128.1
14
3167
XM_011537160.2
XP_011535462.1
17
3763
XM_017021648.1
XP_016877137.1
16
3572
GenBank mRNA
AF474376.1
-
7
1508
AL359625.1
-
1
781
AY578063.1
-
9
2188
BX640876.1
-
6
1848
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of mRNA stability
negative regulation of mRNA polyadenylation
protein binding
poly(A) binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0