Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZC3H14 SIRNA Assays ›
Gene Symbol:
ZC3H14
Gene Name:
zinc finger CCCH-type containing 14
Gene Aliases:
MSUT-2, NY-REN-37, SUT2, UKp68
Chromosome Location:
Chr.14: 88562909 - 88614720 on Build GRCh38
UniGene ID:
Hs.684046
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
258618
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZC3H14
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001160103.1
NP_001153575.1
17
2979
NM_001160104.1
NP_001153576.1
17
2964
NM_001326295.1
NP_001313224.1
15
2589
NM_001326296.1
NP_001313225.1
16
2904
NM_001326297.1
NP_001313226.1
17
3163
NM_001326298.1
NP_001313227.1
14
2514
NM_001326299.1
NP_001313228.1
16
2889
NM_001326300.1
NP_001313229.1
16
2867
NM_001326301.1
NP_001313230.1
16
3088
NM_001326302.1
NP_001313231.1
16
2864
NM_001326303.1
NP_001313232.1
14
2692
NM_001326304.1
NP_001313233.1
14
2471
NM_001326305.1
NP_001313234.1
15
2792
NM_001326306.1
NP_001313235.1
13
2355
NM_001326307.1
NP_001313236.1
16
2907
NM_001326308.1
NP_001313237.1
14
2474
NM_001326309.1
NP_001313238.1
13
2399
NM_001326310.1
NP_001313239.1
17
2967
NM_001326311.1
NP_001313240.1
13
2396
NM_001326312.1
NP_001313241.1
16
2892
NM_001326313.1
NP_001313242.1
13
2358
NM_001326314.1
NP_001313243.1
15
2789
NM_001326315.1
NP_001313244.1
17
3160
NM_001326316.1
NP_001313245.1
17
2827
NM_024824.4
NP_079100.2
17
2982
NM_207660.3
NP_997543.1
14
2511
NM_207661.2
NP_997544.1
15
2437
NM_207662.3
NP_997545.2
7
1808
NR_136936.1
16
2822
XM_005268068.4
XP_005268125.1
16
2902
XM_005268069.4
XP_005268126.1
15
2584
XM_005268070.4
XP_005268127.1
15
2581
XM_005268071.4
XP_005268128.1
14
2494
XM_011537160.2
XP_011535462.1
17
3090
XM_017021648.1
XP_016877137.1
16
2899
GenBank mRNA
AK090809.1
-
3
2042
AL359625.1
-
1
108
AY578061.1
-
14
2450
AY578063.1
-
9
1515
BX248769.1
-
4
872
BX640876.1
-
6
1175
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of mRNA stability
negative regulation of mRNA polyadenylation
protein binding
poly(A) binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0