Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipmk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027184.1 | NP_081460.1 | 6 | 2412 | ||
GenBank mRNA | AK034274.1 | - | 1 | 515 | ||
AK036084.1 | - | 6 | 2180 | |||
AK039460.1 | - | 1 | 1356 | |||
BC052463.1 | - | 6 | 2412 | 6834748 |