Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027551.2 | NP_081827.1 | 8 | 2982 | ||
GenBank mRNA | AK014532.1 | - | 4 | 1849 | ||
AK086025.1 | - | 8 | 2983 | |||
AK146580.1 | - | 8 | 3039 | |||
AK148398.1 | - | 8 | 2992 | |||
AK185832.1 | - | 1 | 162 | |||
AK190841.1 | - | 1 | 162 | |||
AK199574.1 | - | 1 | 163 | |||
AK205662.1 | - | 2 | 346 |