Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS37C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017966.4 | NP_060436.4 | 5 | 2134 | ||
XM_005274077.3 | XP_005274134.1 | 5 | 2175 | |||
XM_017017955.1 | XP_016873444.1 | 2 | 1844 | |||
GenBank mRNA | AK000854.1 | - | 1 | 722 | ||
AK302384.1 | - | 6 | 1154 | |||
AL834261.1 | - | 5 | 2142 | |||
BC005805.2 | - | 1 | 761 | |||
BC062604.1 | - | 1 | 857 | |||
BC141827.1 | - | 5 | 2097 | 40148285 | ||
BC142702.1 | - | 5 | 2097 | 40148798 | ||
CR457250.1 | - | 1 | 401 |