Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168297.1 | NP_001161769.1 | 3 | 3215 | ||
NM_027968.3 | NP_082244.2 | 3 | 3249 | |||
GenBank mRNA | AK047174.1 | - | 1 | 847 | ||
AK083479.1 | - | 3 | 3206 | |||
AK135274.1 | - | 3 | 3208 | |||
AK137546.1 | - | 1 | 262 |