Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029153.1 | NP_083429.1 | 9 | 2606 | ||
GenBank mRNA | AK135058.1 | - | 9 | 2620 | ||
AK154671.1 | - | 9 | 2625 | |||
BC034283.1 | - | 9 | 2606 | 4162134 |