Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083614.1 | NP_001077083.1 | 9 | 1772 | ||
NR_003501.1 | 9 | 1772 | ||||
XM_011545738.1 | XP_011544040.1 | 9 | 2062 | |||
XM_011545739.1 | XP_011544041.1 | 9 | 1681 | |||
GenBank mRNA | AB075850.1 | - | 9 | 1729 | ||
AK095998.1 | - | 9 | 1761 | |||
AL832489.1 | - | 9 | 1748 | |||
BX648129.1 | - | 7 | 1451 |