Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013666.2 | NP_038694.2 | 8 | 1926 | ||
NM_153124.2 | NP_694764.2 | 7 | 1818 | |||
GenBank mRNA | AK013542.1 | - | 7 | 1815 | ||
AK078670.1 | - | 7 | 1602 | |||
AK162549.1 | - | 7 | 1598 | |||
BC034855.1 | - | 7 | 1812 | 5146703 | ||
X98014.1 | - | 8 | 1789 |