Search Thermo Fisher Scientific
Gene Symbol: KGFLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003674.2 | 1 | 146 | |||
NR_047527.1 | 1 | 146 | ||||
GenBank mRNA | AF523265.1 | - | 1 | 241 | ||
AY098593.1 | - | 1 | 137 | |||
BC101842.1 | - | 1 | 146 | |||
BC112977.1 | - | 1 | 144 | |||
BC112978.1 | - | 1 | 144 |
Gene Symbol: KGFLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003670.1 | 1 | 245 | |||
GenBank mRNA | AY098594.1 | - | 1 | 138 |
Gene Symbol: LOC100126582 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK296330.1 | - | 1 | 153 | ||
BC108270.1 | - | 1 | 242 | 4776745 |
Gene Symbol: LOC102724238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK290223.1 | - | 1 | 153 |
Gene Symbol: LOC403323 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122076.1 | 1 | 257 | |||
NR_122077.1 | 1 | 257 | ||||
GenBank mRNA | AK000451.1 | - | 1 | 257 |