Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318781.1 | NP_001305710.1 | 22 | 3331 | ||
NM_182920.1 | NP_891550.1 | 23 | 3415 | |||
GenBank mRNA | AB037733.1 | - | 22 | 2910 | ||
AF488803.1 | - | 23 | 3415 | |||
AK300357.1 | - | 2 | 87 | |||
AK303343.1 | - | 4 | 550 | |||
BC130578.1 | - | 23 | 3405 | 40146609 | ||
BC171764.1 | - | 22 | 3321 | 9054417 |