Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HRASLS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017878.1 | NP_060348.1 | 4 | 596 | ||
XM_011545120.2 | XP_011543422.1 | 5 | 1048 | |||
GenBank mRNA | AK000563.1 | - | 4 | 596 |