Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Riok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009687.1 | NP_001009687.1 | 6 | 747 | ||
GenBank mRNA | BC088297.1 | - | 6 | 747 | 7322759 |