Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xkr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011732.1 | NP_001011732.1 | 3 | 2038 | ||
GenBank mRNA | AY534253.1 | - | 3 | 2038 |