Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM118A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104595.1 | NP_001098065.1 | 10 | 1690 | ||
NM_017911.2 | NP_060381.2 | 9 | 1363 | |||
XM_011530247.2 | XP_011528549.1 | 9 | 1481 | |||
XM_011530254.2 | XP_011528556.1 | 9 | 1271 | |||
XM_017028841.1 | XP_016884330.1 | 9 | 1266 | |||
XM_017028842.1 | XP_016884331.1 | 8 | 1033 | |||
XM_017028843.1 | XP_016884332.1 | 8 | 1030 | |||
XM_017028844.1 | XP_016884333.1 | 7 | 1000 | |||
XM_017028845.1 | XP_016884334.1 | 10 | 1466 | |||
XM_017028846.1 | XP_016884335.1 | 9 | 1350 | |||
XM_017028847.1 | XP_016884336.1 | 9 | 1994 | |||
XM_017028848.1 | XP_016884337.1 | 9 | 1392 | |||
XM_017028849.1 | XP_016884338.1 | 9 | 1228 | |||
XM_017028850.1 | XP_016884339.1 | 9 | 1351 | |||
XM_017028851.1 | XP_016884340.1 | 9 | 1982 | |||
XM_017028852.1 | XP_016884341.1 | 9 | 1388 | |||
XM_017028854.1 | XP_016884343.1 | 8 | 1115 | |||
XM_017028861.1 | XP_016884350.1 | 9 | 1665 | |||
GenBank mRNA | AK000642.1 | - | 9 | 1333 | ||
AK125027.1 | - | 10 | 1908 | |||
AK302205.1 | - | 6 | 842 | |||
BC013696.1 | - | 9 | 1120 | 3860694 | ||
CB961784.1 | - | 6 | 653 | |||
CR456403.1 | - | 8 | 1089 |