Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012038.1 | NP_001012038.1 | 16 | 3176 | ||
XM_006251302.2 | XP_006251364.1 | 18 | 3315 | |||
XM_006251304.2 | XP_006251366.1 | 19 | 3371 | |||
XM_006251305.2 | XP_006251367.1 | 17 | 3179 | |||
XM_008770298.1 | XP_008768520.1 | 19 | 4494 | |||
XM_008770299.1 | XP_008768521.1 | 19 | 3381 | |||
XM_008770300.1 | XP_008768522.1 | 18 | 3284 | |||
XM_008770301.1 | XP_008768523.1 | 16 | 3094 | |||
XM_008770302.1 | XP_008768524.1 | 19 | 3369 | |||
XM_008770303.1 | XP_008768525.1 | 19 | 3369 | |||
XM_008770304.1 | XP_008768526.1 | 19 | 3369 | |||
XM_008770305.1 | XP_008768527.1 | 19 | 3369 | |||
XM_008770306.1 | XP_008768528.1 | 19 | 3369 | |||
GenBank mRNA | BC087045.1 | - | 16 | 3176 | 7109341 |