Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Samd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012040.1 | NP_001012040.1 | 3, 4 | 907 | ||
XM_006251640.1 | XP_006251702.1 | 3, 4 | 945 | |||
GenBank mRNA | BC082100.1 | - | 3, 4 | 907 | 7134467 | |
FQ219794.1 | - | 3, 4 | 945 |