Search Thermo Fisher Scientific
Gene Symbol: Btd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012047.1 | NP_001012047.1 | 3 | 483 | ||
XM_006252609.2 | XP_006252671.1 | 5 | 618 | |||
XM_006252610.1 | XP_006252672.1 | 4 | 503 | |||
XM_006252611.2 | XP_006252673.1 | 4 | 1697 | |||
XM_006252612.1 | XP_006252674.1 | 6 | 1008 | |||
XM_006252614.1 | XP_006252676.1 | 5 | 885 | |||
XM_006252615.2 | XP_006252677.1 | 4 | 833 | |||
XM_006252616.2 | XP_006252678.1 | 4 | 1614 | |||
XM_006252617.1 | XP_006252679.1 | 4 | 737 | |||
XM_006252618.1 | XP_006252680.1 | 3 | 470 | |||
XM_006252619.2 | XP_006252681.1 | 6 | 645 | |||
XM_006252620.2 | XP_006252682.1 | 5 | 659 | |||
XM_006252622.2 | XP_006252684.1 | 4 | 614 | |||
XM_008770998.1 | XP_008769220.1 | 4 | 457 | |||
XM_008770999.1 | XP_008769221.1 | 4 | 774 | |||
GenBank mRNA | BC090017.1 | - | 3 | 483 | 7325227 |