Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tada2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012141.1 | NP_001012141.1 | 12 | 1043 | ||
XM_006247041.2 | XP_006247103.1 | 11 | 1006 | |||
XM_006247042.2 | XP_006247104.1 | 11 | 952 | |||
GenBank mRNA | BC079084.1 | - | 12 | 1043 | 7123007 | |
EF121983.1 | - | 11 | 882 | |||
EF121984.1 | - | 11 | 882 | |||
EF121985.1 | - | 10 | 809 |