Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012970.2 | NP_001012988.1 | 5 | 566 | ||
NM_001276348.1 | NP_001263277.1 | 5 | 470 | |||
NM_001276349.1 | NP_001263278.1 | 4 | 393 | |||
XM_011544124.2 | XP_011542426.1 | 6 | 1408 | |||
XM_011544125.2 | XP_011542427.1 | 5 | 1158 | |||
XM_011544128.2 | XP_011542430.1 | 5 | 1669 | |||
XM_011544129.2 | XP_011542431.1 | 6 | 1319 | |||
XM_011544130.2 | XP_011542432.1 | 5 | 1240 | |||
XM_017000571.1 | XP_016856060.1 | 6 | 1438 | |||
XM_017000572.1 | XP_016856061.1 | 6 | 1946 | |||
GenBank mRNA | BC054479.1 | - | 5 | 474 | ||
BX099227.1 | - | 4 | 379 |