Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013036.1 | NP_001013054.1 | 10 | 1478 | ||
GenBank mRNA | BC089848.1 | - | 10 | 1478 | 7374684 | |
FQ211507.1 | - | 11 | 2390 |