Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lman2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013374.1 | NP_001013392.1 | 5, 6 | 670 | ||
GenBank mRNA | AK041656.1 | - | 3, 4 | 2515 | ||
AK142024.1 | - | 639 | ||||
AK158015.1 | - | 5, 6 | 689 | |||
BC027226.1 | - | 4, 5 | 588 | 2654285 | ||
BC046969.1 | - | 5, 6 | 670 | 6313679 |