Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011534534.2 | XP_011532836.1 | 4 | 974 | ||
XM_011534535.1 | XP_011532837.1 | 4 | 1159 | |||
GenBank mRNA | AK093042.1 | - | 1 | 517 |
Gene Symbol: LOC100289470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093197.1 | - | 2 | 1766 |